missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nipecotamide, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 128300050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nipecotamide | |
4138-26-5 | |
100.0 | |
95% | |
C6H12N2O | |
5g | |
BVOCPVIXARZNQN-UHFFFAOYSA-N | |
piperidine-3-carboxamide | |
92980 | |
128.17 |
95% | |
94.0 | |
Authentic | |
Glass bottle | |
MFCD00005993 | |
nipecotamide, 3-piperidinecarboxamide, nipecotic acid amide, hexahydronicotinamide, piperidine-3-carboxylic acid amide, racemic nipecotamide, pubchem11797, acmc-20lak2, 3-piperidine carboxamide | |
C1CC(CNC1)C(=O)N | |
128.17 | |
CHEBI:60116 | |
95% |
Chemical Identifiers
4138-26-5 | |
128.17 | |
BVOCPVIXARZNQN-UHFFFAOYSA-N | |
92980 | |
piperidine-3-carboxamide |
C6H12N2O | |
MFCD00005993 | |
nipecotamide, 3-piperidinecarboxamide, nipecotic acid amide, hexahydronicotinamide, piperidine-3-carboxylic acid amide, racemic nipecotamide, pubchem11797, acmc-20lak2, 3-piperidine carboxamide | |
CHEBI:60116 | |
C1CC(CNC1)C(=O)N |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
EINECSNumber : 223-962-5
RUO â Research Use Only