missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium Wire, 1.0mm (0.04 in.) dia., 0.998, Alfa Aesar™

Form: Approx.6.7g/m

Manufacturer:  Thermo Fisher Scientific 10264G5

 View more versions of this product

Catalog No. AA10264G5



Specifications

Specifications

Niobium
7440-03-1
99.80%
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
92.91
CHEBI:33344
Silver to Gray
5m
Silver gray wire
Nb
columbio
[Nb]
Odorless
23936
Wire, 1.0mm (0.04 in.) dia.

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbio
CHEBI:33344
[Nb]
Documents