missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Niobium wire, 0.25mm (0.01in) dia, 99.99% (metals basis excluding Ta), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 010265CC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Niobium wire | |
7440-03-1 | |
Nb | |
500 cm | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
niobium | |
23936 | |
99.99% | |
≈0.42g/m |
0.25mm (0.01 in.) dia. | |
(metals basis excluding Ta) | |
MFCD00011126 | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
[Nb] | |
92.91 | |
CHEBI:33344 | |
Odorless | |
Wire |
Chemical Identifiers
7440-03-1 | |
92.91 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
23936 | |
niobium |
Nb | |
MFCD00011126 | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
CHEBI:33344 | |
[Nb] |
Safety and Handling
EINECSNumber : 231-113-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only