missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium wire, 0.25mm (0.01 in.) dia., 99.99% (metals basis excluding Ta)

Manufacturer:  Alfa Aesar™ 10265BU

 View more versions of this product

Catalog No. AA10265BU



Specifications

Specifications

Niobium wire
25cm
7440-03-1
Nb
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
[Nb]
Odorless
23936
Wire
≈0.42g/m
0.25mm (0.01 in.) dia.
(metals basis excluding Ta)
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
92.91
CHEBI:33344
99.99%

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
CHEBI:33344
[Nb]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents