missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium(V) oxide, 99.9985% (metals basis), Alfa Aesar™ Puratronic™

Manufacturer:  Thermo Fisher Scientific 1082218

 View more versions of this product

Catalog No. AA1082218



Specifications

Specifications

Niobium(V) oxide
99.9985% (metals basis)
MFCD00011128
diniobium pentaoxide
O=[Nb](=O)O[Nb](=O)=O
265.808
265.81
4.6g/cm3 at 20°C
White
1313-96-8
Nb2O5
White powder or lump
ZKATWMILCYLAPD-UHFFFAOYSA-N
dioxoniobiooxy(dioxo)niobium
123105
Powder
Odorless
50g

Chemical Identifiers

1313-96-8
265.808
ZKATWMILCYLAPD-UHFFFAOYSA-N
123105
O=[Nb](=O)O[Nb](=O)=O
Nb2O5
MFCD00011128
diniobium pentaoxide
dioxoniobiooxy(dioxo)niobium
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-213-6

Documents