Please login to your online account to display your discounted pricing

Niobium(V) oxide, 99.5% (metals basis), Alfa Aesar™

As an intermediate and in electronics

Chemical Identifiers

Chemical Name or Material Niobium(V) oxide
CAS 1313-96-8
Molecular Formula Nb2O5
Formula Weight 265.81
MDL Number MFCD00011128
Merck Index 14,6559
Synonym Niobium pentoxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1136536

  SDS
alfa aesar
1136536
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 265.81
Quantity 500g
Chemical Name or Material Niobium(V) oxide
TSCA TSCA
Mesh Size -100 Mesh
Odor Odorless
Physical Form Powder
Assay 99.5%
Molecular Formula Nb2O5
MDL Number MFCD00011128
CAS 1313-96-8
Merck Index 14,6559
Synonym Niobium pentoxide
Melting Point 1520°C
Solubility Information Insoluble in water. Soluble in HF and alkalies
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.6
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.