missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium(V) fluoride, 99%

Manufacturer:  Alfa Aesar™ 8722109

 View more versions of this product

Catalog No. AA8722109



Specifications

Specifications

Niobium (V) fluoride
NbF5
UN3260
Niobium pentafluoride
187.91
99%
Moisture Sensitive
White to Yellow
10g
7783-68-8
MFCD00011129
14,6558
Hydrolyzes in water, alcohol, and caustic solutions; Sparingly soluble in CS2, chloroform; Forms complexes with Lewis bases
Powder
3.92g/mL
~230°C
80°C

Chemical Identifiers

7783-68-8
NbF5
MFCD00011129
Niobium pentafluoride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents