missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium(V) ethoxide, 99.999% (metals basis), Ta <500ppm

Preparation of niobium oxide and mixed metal oxides

Manufacturer:  Alfa Aesar™ 1468909

 View more versions of this product

Catalog No. AA1468909



Specifications

Specifications

Niobium(V) ethoxide
36° (97°F)
Moisture Sensitive
6°C
3236-82-6
C10H25NbO5
MFCD00015122
3678883
Ta <500ppm
ZTILUDNICMILKJ-UHFFFAOYSA-N
ethanolate;niobium(5+)
160675
Liquid
1.258g/mL
1.5160
140°C to 142°C (0.1mmHg)
10g
(metals basis)
Nb(OC2H5)5
UN2924
niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
Soluble in organic solvents; Decomposes in water
CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]
318.211
318.21
99.999%

Chemical Identifiers

3236-82-6
318.211
ZTILUDNICMILKJ-UHFFFAOYSA-N
160675
CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].[Nb+5]
C10H25NbO5
MFCD00015122
niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
ethanolate;niobium(5+)
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

SDS Documents