Please login to your online account to display your discounted pricing

Niobium(V) ethoxide, 99.95% (trace metal basis), ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC316880500

 View more versions of this product

Catalog No. AC316880500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Liquid
Formula Weight 318.21
Quantity 50g
Color Yellow to orange-brown
Density 1.2680g/mL
CAS Max % 100.0
Packaging Glass bottle
Linear Formula Nb(OC2H5)5
Total Trace Metal Impurities 500ppm max.
CAS 3236-82-6
Chemical Name or Material Niobium(V) ethoxide
Molecular Formula C10H25NbO5
Assay Percent Range 99.95%
MDL Number MFCD00015122
Health Hazard 2 GHS P Statement Flammable liquid and vapor. Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co
Health Hazard 1 Danger
CAS Min % 99.9

Specifications

Chemical Name or Material Niobium(V) ethoxide
CAS 3236-82-6
CAS Min % 99.9
CAS Max % 100.0
Purity Grade Trace Metal Basis
Assay Percent Range 99.95%
Molecular Formula C10H25NbO5
Linear Formula Nb(OC2H5)5
Formula Weight 318.21
MDL Number MFCD00015122
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Flammable liquid and vapor. Causes severe skin burns and eye damage.

SDS & Safety Documents