missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium(V) chloride, Alfa Aesar™

Manufacturer:  Alfa Aesar 1154814

 View more versions of this product

Catalog No. AA1154814



Specifications

Specifications

Niobium(V) chloride
99.9% (metals basis)
MFCD00011127
columbium pentachloride
Cl[Nb](Cl)(Cl)(Cl)Cl
270.156
270.2
Acidic
25g
10026-12-7
Cl5Nb
Yellow powder/flake/crystalline/beads
YHBDIEWMOMLKOO-UHFFFAOYSA-I
pentachloroniobium
24818
Powder, Flake, Crystalline, or Beads
White

Chemical Identifiers

10026-12-7
270.156
YHBDIEWMOMLKOO-UHFFFAOYSA-I
24818
Cl[Nb](Cl)(Cl)(Cl)Cl
Cl5Nb
MFCD00011127
columbium pentachloride
pentachloroniobium
Safety and Handling

Safety and Handling

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 233-059-8

Documents