missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium(V) chloride, Alfa Aesar™

$97.78 - $902.14

Chemical Identifiers

CAS 10026-12-7
Molecular Formula Cl5Nb
Molecular Weight (g/mol) 270.156
MDL Number MFCD00011127
InChI Key YHBDIEWMOMLKOO-UHFFFAOYSA-I
Synonym columbium pentachloride
PubChem CID 24818
IUPAC Name pentachloroniobium
SMILES Cl[Nb](Cl)(Cl)(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1154814 View Documents Alfa Aesar
1154814
25g Each for $97.78
 
AA1154822 View Documents Alfa Aesar
1154822
100g Each for $208.19
 
AA1154836 View Documents Alfa Aesar
1154836
500g Each for $902.14
 
Specifications

Chemical Identifiers

10026-12-7
270.156
YHBDIEWMOMLKOO-UHFFFAOYSA-I
24818
Cl[Nb](Cl)(Cl)(Cl)Cl
Cl5Nb
MFCD00011127
columbium pentachloride
pentachloroniobium

Specifications

10026-12-7
Cl5Nb
Yellow powder/flake/crystalline/beads
YHBDIEWMOMLKOO-UHFFFAOYSA-I
pentachloroniobium
24818
Powder, Flake, Crystalline, or Beads
White
Niobium(V) chloride
99.9% (metals basis)
MFCD00011127
columbium pentachloride
Cl[Nb](Cl)(Cl)(Cl)Cl
270.156
270.2
Acidic
25g
Safety and Handling

Safety and Handling

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 233-059-8