missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium(V) chloride, 99% (metals basis), Alfa Aesar™

For Use in Preparation of Pure Niobium, and As An Intermediate

Manufacturer:  Alfa Aesar 5110836

 View more versions of this product

Catalog No. AA5110836



Specifications

Specifications

Niobium(V) chloride
99% (metals basis)
MFCD00011127
YHBDIEWMOMLKOO-UHFFFAOYSA-I
pentachloroniobium
24818
15mm and down Lumps, Packaged Under Argon
Yellow
10026-12-7
Cl5Nb
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, niobium v chloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
Cl[Nb](Cl)(Cl)(Cl)Cl
270.156
270.2
Acidic
500g

Chemical Identifiers

10026-12-7
270.156
YHBDIEWMOMLKOO-UHFFFAOYSA-I
24818
Cl[Nb](Cl)(Cl)(Cl)Cl
Cl5Nb
MFCD00011127
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, niobium v chloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
pentachloroniobium
Safety and Handling

Safety and Handling

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 233-059-8

SDS Documents