missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium standard solution, for AAS, 1mg/mL Nb in 5% HF, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 196161000

 View more versions of this product

Catalog No. AC196161000


 

Specifications Description & Specifications

Specifications

Niobium standard solution
7440-03-1,7664-39-3,7732-18-5
5.1,94.1
Liquid
995 to 1005 μg/mL of Nb in 5% HF (20°C)
MFCD00011126
1.01g/mL
100mL
1 mg/ml Nb in 5% HF
4.9,93.9
For AAS
Nb
92.91
Plastic bottle
Clear colorless

Chemical Identifiers

7440-03-1,7664-39-3,7732-18-5
5.1,94.1
92.91
Niobium standard solution, 1 mg/ml Nb in 5% HF
4.9,93.9
Nb
MFCD00011126
For AAS

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger