Please login to your online account to display your discounted pricing

Niobium standard solution, for AAS, 1mg/mL Nb in 5% HF, ACROS Organics™

$31.00 - $31.00

Chemical Identifiers

Chemical Name or Material Niobium standard solution
Name Note 1 mg/ml Nb in 5% HF
CAS 7440-03-1
CAS 7664-39-3
CAS Min % 4.9
CAS Max % 5.1
CAS 7732-18-5
CAS Min % 93.9
CAS Max % 94.1
Purity Grade For AAS
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC196161000

  SDS
thermo fisher scientific
196161000
Plastic bottle 100mL Each for $31.00
Description & Specifications

Specifications

Purity Grade For AAS
Physical Form Liquid
Formula Weight 92.91
Quantity 100mL
Color Clear colorless
Density 1.01g/mL
Chemical Name or Material Niobium standard solution
CAS Max % 94.1
Name Note 1 mg/ml Nb in 5% HF
Molecular Formula Nb
Concentration 995 to 1005 μg/mL of Nb in 5% HF (20°C)
MDL Number MFCD00011126
CAS 7440-03-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
CAS 7664-39-3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
CAS Min % 4.9
CAS Max % 5.1
CAS 7732-18-5
CAS Min % 93.9

for atomic adsorption spectroscopy

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.