Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis excluding Ta)

$824.20 - $824.20

Chemical Identifiers

Chemical Name or Material Niobium sputtering target
CAS 7440-03-1
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00011126
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40885KS

  SDS
alfa aesar™
40885KS
1 each Each for $824.20
Description & Specifications

Specifications

Chemical Name or Material Niobium sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis excluding Ta)
MDL Number MFCD00011126
CAS 7440-03-1
Odor Odorless
Quantity 1 each