Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis including Ta)

$367.65 - $367.65

Chemical Identifiers

Chemical Name or Material Niobium sputtering target
CAS 7440-03-1
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011126
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40884KS

  SDS
alfa aesar™
40884KS
1 each Each for $367.65
Description & Specifications

Specifications

MDL Number MFCD00011126
Assay 99.95%
Odor Odorless
TSCA TSCA
Chemical Name or Material Niobium sputtering target
Assay Percent Notes (metals basis excluding Ta)
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Quantity 1 each
CAS 7440-03-1