Learn More
Niobium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 040883KS
Description
Niobium sputtering target is used in high temperature engineering products. It is also used as an alloying agent for certain steels where it greatly improves the strength of the resulting material. It is also finds applications in atomic reactors due to its corrosion resistance and, when combined with either tin (Nb3Sn) or zirconium, it has a high degree of superconductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Niobium sputtering target | |
50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick | |
Odorless | |
Nb | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
niobium | |
23936 | |
99.95% |
1 Ea. | |
7440-03-1 | |
(metals basis excluding Ta) | |
MFCD00011126 | |
Insoluble in water. | |
[Nb] | |
92.91 | |
CHEBI:33344 |
Chemical Identifiers
7440-03-1 | |
92.91 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
23936 | |
niobium |
Nb | |
MFCD00011126 | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
CHEBI:33344 | |
[Nb] |
Safety and Handling
EINECSNumber : 231-113-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only