Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis excluding Ta)

Magnetron sputtering source

$536.41 - $536.41

Chemical Identifiers

Chemical Name or Material Niobium sputtering target
CAS 7440-03-1
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011126
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40882KS

  SDS
alfa aesar™
40882KS
1 each Each for $536.41
Description & Specifications

Specifications

Quantity 1 each
Odor Odorless
Chemical Name or Material Niobium sputtering target
Assay Percent Notes (metals basis excluding Ta)
TSCA TSCA
MDL Number MFCD00011126
CAS 7440-03-1
Assay 99.95%
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick