missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium silicide, 99.85% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1310022

 View more versions of this product

Catalog No. AA1310022



Specifications

Specifications

Niobium silicide
99.85% (metals basis)
MFCD00151537
bis $l^ 2
GXAAOBARRLPQRY-UHFFFAOYSA-N
bis($l^{2}-silanylidene)niobium
6336884
-325 Mesh Powder
1950°C
12034-80-9
NbSi2
Powder
Insoluble in water.
[Si]=[Nb]=[Si]
149.076
149.08
5.7
100g

Chemical Identifiers

12034-80-9
149.076
GXAAOBARRLPQRY-UHFFFAOYSA-N
6336884
[Si]=[Nb]=[Si]
NbSi2
MFCD00151537
bis $l^ 2
bis($l^{2}-silanylidene)niobium
Safety and Handling

Safety and Handling

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

EINECSNumber : 234-812-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents