Please login to your online account to display your discounted pricing

Niobium silicide, 99.5% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Niobium silicide
CAS 12060-34-3
Physical Form Powder
Assay Percent Range 99.5% (metals basis)
Molecular Formula Nb5Si3
Formula Weight 548.79
MDL Number MFCD00213811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3947114

  SDS
alfa aesar
3947114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3947122

  SDS
alfa aesar
3947122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 548.79
Quantity 25g
Chemical Name or Material Niobium silicide
Physical Form Powder
Assay Percent Range 99.5% (metals basis)
Molecular Formula Nb5Si3
Appearance Powder
MDL Number MFCD00213811
CAS 12060-34-3
Health Hazard 1 H335: May cause respiratory irritation.
Safety and Handling

H335: May cause respiratory irritation.