missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium rod, 12.7mm (0.5 in.) dia., hard, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 43362BP

 View more versions of this product

Catalog No. AA43362BP



Specifications

Specifications

Niobium rod
2cm
7440-03-1
Nb
columbio
[Nb]
Odorless
23936
Rod
≈10.9g/cm
12.7mm (0.5 in.) dia., Hard, Temper: as drawn
(metals basis)
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
92.91
CHEBI:33344
99.8%

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbio
CHEBI:33344
[Nb]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents