missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium potassium n-butoxide, 5% w/v in n-butanol, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 44209AC

 View more versions of this product

Catalog No. AA44209AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Niobium potassium n-butoxide
25mL
C24H54KNbO6
MFCD02682834
5% w/v in n-butanol
Liquid
Moisture sensitive
(metals basis)
KNb(OC4H9)6
UN1120
570.01
99%

Chemical Identifiers

C24H54KNbO6
MFCD02682834
Safety and Handling

Safety and Handling

GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H318-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS

TSCA : No

Recommended Storage : Ambient temperatures

Documents