Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium potassium n-butoxide, 5% w/v in n-butanol, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Niobium potassium n-butoxide
Molecular Formula C24H54KNbO6
Linear Formula KNb(OC4H9)6
Formula Weight 570.01
MDL Number MFCD02682834
UN Number UN1120
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44209AC

  SDS
alfa aesar™
44209AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA44209AE

  SDS
alfa aesar™
44209AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 570.01
Chemical Name or Material Niobium potassium n-butoxide
Physical Form Liquid
Assay 99%
Assay Percent Notes (metals basis)
Molecular Formula C24H54KNbO6
Linear Formula KNb(OC4H9)6
MDL Number MFCD02682834
UN Number UN1120
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P280d-P301+P310-P305+P351+P338-P420a-P403+P233
Wear protective clothing and eye protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store away from foodstuffs.
Store in a well-ventilated place.
Keep container tightly closed.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
Concentration 5% w/v in n-butanol
Sensitivity Moisture Sensitive
Quantity 25mL
Safety and Handling

Danger

GHS P Statement
P280d-P301+P310-P305+P351+P338-P420a-P403+P233
Wear protective clothing and eye protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store away from foodstuffs.
Store in a well-ventilated place.
Keep container tightly closed.

GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.