missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium potassium n-butoxide, 5% w/v in n-butanol, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 44209AE

 View more versions of this product

Catalog No. AA44209AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Niobium potassium n-butoxide
100mL
C24H54KNbO6
MFCD02682834
5% w/v in n-butanol
Liquid
Moisture Sensitive
(metals basis)
KNb(OC4H9)6
UN1120
570.01
99%

Chemical Identifiers

C24H54KNbO6
MFCD02682834
Safety and Handling

Safety and Handling

GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.

GHS P Statement
P280d-P301+P310-P305+P351+P338-P420a-P403+P233
Wear protective clothing and eye protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store away from foodstuffs.
Store in a well-ventilated place.
Keep container tightly closed.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS

SDS Documents