Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium potassium isopropoxide, 2.5% w/v in isopropanol, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Niobium potassium isopropoxide
CAS 21864-23-3
Linear Formula KNb[OCH(CH3)2]6
Formula Weight 486.53
MDL Number MFCD00798516
UN Number UN1219
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39165AD

  SDS
alfa aesar™
39165AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39165AH

  SDS
alfa aesar™
39165AH
200mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 486.53
Chemical Name or Material Niobium potassium isopropoxide
Physical Form Liquid
Assay 99%
Assay Percent Notes (metals basis)
Linear Formula KNb[OCH(CH3)2]6
MDL Number MFCD00798516
CAS 21864-23-3
UN Number UN1219
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Concentration 2.5% w/v in isopropanol
Flash Point 12° (54°F)
Odor Alcohol-like
Sensitivity Moisture Sensitive
Color Pale yellow to brown
Quantity 50mL
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.