missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium phenoxide

Manufacturer:  Alfa Aesar™ 5112002

 View more versions of this product

Catalog No. AA5112002



Specifications

Specifications

Niobium phenoxide
208°C
16576-63-9
Nb(C6H5O)5
niobium 5+ pentakisphenoxide, pentaphenoxyniobium v, niobium 5+ ion pentaphenolate, columbium penta phenoxide, niobium 5+ pentaphenolate, phenol, niobium 5+ salt 5:1
C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].[Nb+5]
558.431
558.44
Moisture Sensitive
0.5g
C30H25NbO5
MFCD00014076
ZCNDFHXTXLMYDM-UHFFFAOYSA-I
niobium(5+);pentaphenoxide
25021451
Powder

Chemical Identifiers

16576-63-9
558.431
ZCNDFHXTXLMYDM-UHFFFAOYSA-I
25021451
C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].[Nb+5]
C30H25NbO5
MFCD00014076
niobium 5+ pentakisphenoxide, pentaphenoxyniobium v, niobium 5+ ion pentaphenolate, columbium penta phenoxide, niobium 5+ pentaphenolate, phenol, niobium 5+ salt 5:1
niobium(5+);pentaphenoxide
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

SDS Documents