Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium phenoxide

$57.57 - $623.80

Chemical Identifiers

Chemical Name or Material Niobium phenoxide
CAS 16576-63-9
Molecular Formula C30H25NbO5
Linear Formula Nb(C6H5O)5
Formula Weight 558.44
MDL Number MFCD00014076
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA5112002

  SDS
alfa aesar™
5112002
0.5g Each for $57.57

AA5112004

  SDS
alfa aesar™
5112004
2g Each for $168.76

AA5112009

  SDS
alfa aesar™
5112009
10g Each for $623.80
Description & Specifications

Specifications

Formula Weight 558.44
Chemical Name or Material Niobium phenoxide
Physical Form Powder
Molecular Formula C30H25NbO5
Linear Formula Nb(C6H5O)5
MDL Number MFCD00014076
CAS 16576-63-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Sensitivity Moisture Sensitive
Melting Point 208°C
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.