missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium nitride

Manufacturer:  Alfa Aesar™ 1214609

 View more versions of this product

Catalog No. AA1214609



Specifications

Specifications

Niobium nitride
NNb
columbium nitride
N#[Nb]
106.913
106.92
8.40g/mL
2573°C
24621-21-4
MFCD00016239
CFJRGWXELQQLSA-UHFFFAOYSA-N
azanylidyneniobium
90560
Powder
Odorless
10g

Chemical Identifiers

24621-21-4
106.913
CFJRGWXELQQLSA-UHFFFAOYSA-N
90560
N#[Nb]
NNb
MFCD00016239
columbium nitride
azanylidyneniobium
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents