missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium nitride

$84.50 - $296.08

Chemical Identifiers

CAS 24621-21-4
Molecular Formula NNb
Molecular Weight (g/mol) 106.913
MDL Number MFCD00016239
InChI Key CFJRGWXELQQLSA-UHFFFAOYSA-N
Synonym columbium nitride
PubChem CID 90560
IUPAC Name azanylidyneniobium
SMILES N#[Nb]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1214609 View Documents Alfa Aesar™
1214609
10g Each for $84.50
 
AA1214618 View Documents Alfa Aesar™
1214618
50g Each for $296.08
 
Specifications

Chemical Identifiers

24621-21-4
106.913
CFJRGWXELQQLSA-UHFFFAOYSA-N
90560
N#[Nb]
NNb
MFCD00016239
columbium nitride
azanylidyneniobium

Specifications

24621-21-4
MFCD00016239
CFJRGWXELQQLSA-UHFFFAOYSA-N
azanylidyneniobium
90560
Powder
Odorless
10g
NNb
columbium nitride
N#[Nb]
106.913
106.92
8.40g/mL
2573°C
Niobium nitride
Safety and Handling

Safety and Handling

TSCA : TSCA