Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium n-propoxide, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Niobium n-propoxide
CAS 38874-17-8
Linear Formula Nb(OC3H7)5
Formula Weight 388.35
MDL Number MFCD00144917
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3657106

  SDS
alfa aesar™
3657106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3657114

  SDS
alfa aesar™
3657114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.35
Chemical Name or Material Niobium n-propoxide
Physical Form Liquid
Assay 99%
Assay Percent Notes (metals basis)
Linear Formula Nb(OC3H7)5
MDL Number MFCD00144917
CAS 38874-17-8
UN Number UN1993
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Sensitivity Moisture Sensitive
Boiling Point 166°C (0.05mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.