Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium n-pentyloxide, 99% (metals basis)

$50.78 - $760.92

Chemical Identifiers

Chemical Name or Material Niobium n-pentyloxide
CAS 105091-67-6
Linear Formula Nb(OC5H11)5
Formula Weight 528.18
MDL Number MFCD00144918
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3656903

  SDS
alfa aesar™
3656903
1g Each for $50.78

AA3656906

  SDS
alfa aesar™
3656906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3656914

  SDS
alfa aesar™
3656914
25g Each for $760.92
Description & Specifications

Specifications

Formula Weight 528.18
Sensitivity Moisture Sensitive
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Physical Form Liquid
Linear Formula Nb(OC5H11)5
MDL Number MFCD00144918
CAS 105091-67-6
Assay Percent Notes (metals basis)
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 99%
Health Hazard 1 Warning
Chemical Name or Material Niobium n-pentyloxide
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.