Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium isopropoxide, 99% (metals basis), 10% w/v in isopropanol/hexane (50:50)

$158.22 - $158.22

Chemical Identifiers

Chemical Name or Material Niobium isopropoxide
CAS 18368-80-4
Linear Formula Nb[OCH(CH3)2]5
Formula Weight 388.35
MDL Number MFCD00144915
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36572AC

  SDS
alfa aesar™
36572AC
25mL Each for $158.22

AA36572AE

  SDS
alfa aesar™
36572AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.35
MDL Number MFCD00144915
Assay Percent Notes (metals basis)
CAS 18368-80-4
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Moisture Sensitive
Quantity 25mL
UN Number UN1993
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Assay 99%
Chemical Name or Material Niobium isopropoxide
Concentration 10% w/v in isopropanol/hexane (50:50)
Linear Formula Nb[OCH(CH3)2]5
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.