missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium Foil, 1.0mm (0.04 in.) thick, 0.998, Alfa Aesar™

Form: Approx.42.9g/50 x 100mm

Manufacturer:  Thermo Fisher Scientific 10257GK

 View more versions of this product

Catalog No. AA10257GK



Specifications

Specifications

Niobium
7440-03-1
99.80%
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
92.91
CHEBI:33344
Silver to Gray
100 x 200mm
Silver gray foil
Nb
columbio
[Nb]
Odorless
23936
Foil, 1.0mm (0.04 in.) thick

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbio
CHEBI:33344
[Nb]
Documents