missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium foil, 0.5mm (0.02 in.) thick, annealed, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 14358HL

 View more versions of this product

Catalog No. AA14358HL



Specifications

Specifications

Niobium foil
100 x 250mm
7440-03-1
Nb
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
[Nb]
Odorless
23936
Foil
≈21.4g/50x100mm
0.5mm (0.02 in.) thick, Annealed
(metals basis)
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
92.91
CHEBI:33344
99.8%

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
CHEBI:33344
[Nb]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents