missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium Foil, 0.025 ± 0.015mm (0.001 ± 0.0006in) thick, 0.998, Alfa Aesar™

Form: Approx.8.6g/20 x 20cm

$1,834.39 - $1,834.39

Chemical Identifiers

CAS 7440-03-1
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbio, columbium, element, foil, foil, 0.25mm 0.01in thick, niobio, niobium-93, powder, rod, vn 1
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10263CX View Documents Thermo Fisher Scientific
10263CX
20 x 500cm Each for $1,834.39
 
Specifications

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbio, columbium, element, foil, foil, 0.25mm 0.01in thick, niobio, niobium-93, powder, rod, vn 1
CHEBI:33344
[Nb]

Specifications

20 x 500cm
Silver gray foil
Nb
columbio, columbium, element, foil, foil, 0.25mm 0.01in thick, niobio, niobium-93, powder, rod, vn 1
[Nb]
Odorless
23936
Foil, 0.025 ± 0.015mm (0.001 ± 0.0006 in.) thick
Niobium
7440-03-1
99.80%
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
92.91
CHEBI:33344
Silver to Gray