missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium Foil, 0.025 ± 0.015mm (0.001 ± 0.0006in) thick, 0.998, Alfa Aesar™

Form: Approx.8.6g/20 x 20cm

Manufacturer:  Thermo Fisher Scientific 10263CX

 View more versions of this product

Catalog No. AA10263CX



Specifications

Specifications

Niobium
7440-03-1
99.80%
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
92.91
CHEBI:33344
Silver to Gray
20 x 500cm
Silver gray foil
Nb
columbio
[Nb]
Odorless
23936
Foil, 0.025 ± 0.015mm (0.001 ± 0.0006 in.) thick

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbio
CHEBI:33344
[Nb]
Documents