missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Niobium carbide, >99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1214722

 View more versions of this product

Catalog No. AA1214722



Specifications

Specifications

Niobium carbide
99+% (metals basis)
MFCD00016237
niobium carbide nbc, methane-niobium 1:1, methane niobium, niobium carbide trace metals basis 25g
C.[Nb]
108.949
104.91
Odorless
100g
12069-94-2
CH4Nb
Gray powder
CLOIETWQBJYMFR-UHFFFAOYSA-N
methane;niobium
159431
Powder
White

Chemical Identifiers

12069-94-2
108.949
CLOIETWQBJYMFR-UHFFFAOYSA-N
159431
C.[Nb]
CH4Nb
MFCD00016237
niobium carbide nbc, methane-niobium 1:1, methane niobium, niobium carbide trace metals basis 25g
methane;niobium
Safety and Handling

Safety and Handling

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 235-117-8

Documents