Please login to your online account to display your discounted pricing

Niobium carbide, >99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Niobium carbide
CAS 12069-94-2
Physical Form Powder
Assay Percent Range 99+% (metals basis)
Molecular Formula NbC
Formula Weight 104.91
MDL Number MFCD00016237
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1214714

  SDS
alfa aesar
1214714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1214722

  SDS
alfa aesar
1214722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 104.91
Quantity 25g
Color White
DOT Information DOT Class: 4.1, Packaging Group: III
Density 7.6g/mL
Chemical Name or Material Niobium carbide
EINECS Number 235-117-8
Odor Odorless
Physical Form Powder
Assay Percent Range 99+% (metals basis)
Molecular Formula NbC
Appearance Gray powder
MDL Number MFCD00016237
CAS 12069-94-2
Health Hazard 1 H228: Flammable solid.
Safety and Handling

H228: Flammable solid.