missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium boride, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1257314

 View more versions of this product

Catalog No. AA1257314



Specifications

Specifications

Niobium boride
(metals basis)
MFCD00016236
VDZMENNHPJNJPP-UHFFFAOYSA-N
boranylidyneniobium
82882
Powder
Odorless
12007-29-3
BNb
niobium boride, niobium boride nbb, niobiumylidyneborane, niobium monoboride, niobium boride nbb, columbium boride, columbium boride, cl, niobium monoboride
B#[Nb]
103.716
114.53
99%
25g

Chemical Identifiers

12007-29-3
103.716
VDZMENNHPJNJPP-UHFFFAOYSA-N
82882
B#[Nb]
BNb
MFCD00016236
niobium boride, niobium boride nbb, niobiumylidyneborane, niobium monoboride, niobium boride nbb, columbium boride, columbium boride, cl, niobium monoboride
boranylidyneniobium
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents