Please login to your online account to display your discounted pricing

Alfa Aesar™ Niobium boride, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Niobium boride
CAS 12007-29-3
Formula Weight 114.53
MDL Number MFCD00016236
Synonym Niobium diboride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1257314

 
alfa aesar™
1257314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1257322

 
alfa aesar™
1257322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.53
Chemical Name or Material Niobium boride
TSCA TSCA
Odor Odorless
Physical Form Powder
Assay 99%
Assay Percent Notes (metals basis)
MDL Number MFCD00016236
CAS 12007-29-3
Synonym Niobium diboride
Quantity 25g