Please login to your online account to display your discounted pricing

Niobium AA Standard, 1mL = 1mg Nb (1,000ppm Nb)NH4NbF6 in Water / tr HF, Ricca Chemical

$157.76 - $157.76

Chemical Identifiers

Chemical Name or Material Niobium AA Standard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 12062-13-4
CAS Min % 0.25
CAS Max % 0.27
CAS 7664-39-3
CAS Min % 0.1
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ANB1KW500

 
ricca chemical company
RIANB1KW500
Poly natural 500mL Each for $157.76

ANB1KW100

 
ricca chemical company
RIANB1KW100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Odor Odorless
Quantity 500mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg Nb (1, 000ppm Nb)NH4NbF6 in Water / tr HF
Chemical Name or Material Niobium AA Standard
Shelf Life 24 months
CAS Max % 0.1
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 12062-13-4
Packaging Poly natural
CAS Min % 0.25
CAS Max % 0.27
CAS 7664-39-3
CAS Min % 0.1
Safety and Handling

24 months