missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ninhydrin, Monohydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$162.71 - $207.53

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H4O3·H2O
Formula Weight 178.14
Chemical Name or Material Ninhydrin, Monohydrate
Grade Baker Analyzed™ ACS Reagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
02003522 Avantor Performance
N86202
Wide Mouth Amber Glass Bottle 10g Each for $162.71
02003523 Avantor Performance
N86203
Wide Mouth Amber Glass Bottle 25g Each for $207.53
Specifications Description & Specifications

Chemical Identifiers

485-47-2
178.14
Baker Analyzed™ ACS Reagent
C9H4O3·H2O
Ninhydrin, Monohydrate

Specifications

10g
Wide Mouth Amber Glass Bottle
178.14
Baker Analyzed™ ACS Reagent
485-47-2
C9H4O3·H2O
Ninhydrin, Monohydrate