missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nimodipine, 98+%, Thermo Scientific Chemicals
Potent L-type calcium channel antagonist
Supplier: Thermo Scientific Chemicals J61287ME
Description
Potent L-type calcium channel antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nimodipine | |
Yellow | |
MFCD00153848 | |
14,6551 | |
UIAGMCDKSXEBJQ-UHFFFAOYNA-N | |
3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate | |
4497 | |
418.44 | |
Powder |
66085-59-4 | |
C21H26N2O7 | |
500 mg | |
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate | |
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C | |
418.45 | |
CHEBI:7575 | |
≥98% |
Chemical Identifiers
66085-59-4 | |
418.45 | |
UIAGMCDKSXEBJQ-UHFFFAOYNA-N | |
4497 | |
3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
C21H26N2O7 | |
MFCD00153848 | |
nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate | |
CHEBI:7575 | |
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C |
Safety and Handling
GHS H Statement
H312-H332
Harmful in contact with skin.
Harmful if inhaled.
P260-P314-P501c
H373
EINECSNumber : 266-127-0
RTECSNumber : US7975500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only