Nilfisk™ GM 80CR Cleanroom Vacuum
Lightweight, portable and quiet, the GM 80CR is perfect for use in cleanrooms, research labs and other critical environments. Four-stage filtration system includes a 2 1/4-gallon paper bag, main filter, microfilter and ULPA exhaust filter. A conductive hose and accessories provide ESD protection.
Manufacturer: Nilfisk™ 01790132
Description
- 2.25-gallon capacity disposable paper bag captures the bulk of collected debris
- Extra-large main filter ensures a steady, even airflow which extends filter life and eliminates premature clogging
- Conductive plastic hose provides ESD protection
- Wheeled trolley adds mobility
- Positive twist safety latches offer security by preventing the container from opening if the vacuum is dropped
- Vacuum cleaner and accessories are packaged in cleanroom compatible polyethylene bags to prevent contamination from packing materials during shipping
Specifications
Trolley, 1.98m Plastic hose, Steel wands, Paper bags, round brush, crevice cone, nozzles, 9m cord, HEPA filter | |
12.3L | |
Vacuum cleaner | |
GM80CR | |
11.02 lb. | |
409.6 oz., 12.3L |
87 cfm | |
409.6 oz. | |
HEPA | |
120V | |
5kg |