Please login to your online account to display your discounted pricing

Nile Blue A Sulfate, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  15264505

 View more versions of this product

Catalog No. ICN15264505

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 415.47
Color Dark blue
Quantity 5g
EINECS Number 222-832-5
Physical State Solid
MDL Number MFCD00064529
Synonym Basic Blue 12
Recommended Storage Store at Room Temperature (15° to 30°C)
Chemical Name or Material Nile Blue A Sulfate
Molecular Formula C20H20N3O.H2SO4
CAS 2381-85-3

Specifications

Chemical Name or Material Nile Blue A Sulfate
CAS 2381-85-3
Physical Form Powder
Molecular Formula C20H20N3O.H2SO4
Formula Weight 415.47
MDL Number MFCD00064529
Synonym Basic Blue 12
Accessories

Store at Room Temperature (15° to 30°C)