missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nile Blue A Sulfate, MP Biomedicals™

$54.41 - $188.22

Chemical Identifiers

CAS 2381-85-3
Physical Form Powder
Molecular Formula C20H20N3O.H2SO4
Formula Weight 415.47
MDL Number MFCD00064529
Synonym Basic Blue 12
Chemical Name or Material Nile Blue A Sulfate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15264505 Mp Biomedicals
15264505
5g Each for $54.41
ICN15264525 Mp Biomedicals
15264525
25g Each for $188.22
Specifications Description & Specifications

Chemical Identifiers

2381-85-3
C20H20N3O.H2SO4
MFCD00064529
Nile Blue A Sulfate
Powder
415.47
Basic Blue 12

Specifications

2381-85-3
C20H20N3O.H2SO4
MFCD00064529
Solid
5g
Powder
415.47
Basic Blue 12
Dark blue
Nile Blue A Sulfate

Safety and Handling

EINECSNumber : 222-832-5

Recommended Storage : Store at Room Temperature (15° to 30°C)