Please login to your online account to display your discounted pricing

Alfa Aesar™ Nigrosin, soluble in spirit

$27.43 - $54.85

Chemical Identifiers

Chemical Name or Material Nigrosin
CAS 11099-03-9
MDL Number MFCD00149005
Synonym C.I. 50415; Solvent Black 5
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2142414

  SDS
alfa aesar™
B2142414
25g Each for $27.43

AAB2142422

  SDS
alfa aesar™
B2142422
100g Each for $54.85
Description & Specifications

Specifications

TSCA TSCA
Synonym C.I. 50415; Solvent Black 5
Solubility Information Soluble in spirit
RTECS Number GE5800000
Chemical Name or Material Nigrosin
Melting Point >300°C
MDL Number MFCD00149005
Quantity 25g
CAS 11099-03-9