missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nigericin sodium salt, 98+%

A potassium ionophore

$209.73 - $209.73

Chemical Identifiers

CAS 28380-24-7
Molecular Formula C40H68O11
Molecular Weight (g/mol) 724.973
MDL Number MFCD00036825
InChI Key DANUORFCFTYTSZ-FBMOGUJMSA-N
Synonym nigericin
PubChem CID 123134611
IUPAC Name (2R)-2-[(2R,3S,6R)-6-[[(2S,4R,5R,6R,7R,9R)-2-[(2R,5S)-5-[(3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]-5-methyloxolan-2-yl]-7-methoxy-2,4,6-trimethyl-1,10-dioxaspiro[4.5]decan-9-yl]methyl]-3-methyloxan-2-
SMILES CC1CCC(OC1C(C)C(=O)O)CC2CC(C(C3(O2)C(CC(O3)(C)C4CCC(O4)(C)C5C(CC(O5)C6C(CC(C(O6)(CO)O)C)C)C)C)C)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61349MA View Documents Alfa Aesar™
J61349MA
10mg Each for $209.73
 
Description

Description

Applications
Nigericin sodium salt is a polyether ionophore which catalyzes the electroneutral exchange of alkali metal (K+) for H+ (antiport). It disrupts membrane potential and stimulates ATPase activity in mitochondria. It transports monovalent cations across membranes with the following specificity: K+ >Rb+ >Cs+ >>Na. It kills bacteria by facilitating the diffusion of ions across membranes. It is widely used in studies of the consequences of changes in membrane potential in variable systems.

Solubility
Soluble in chloroform, methanol or ethanol

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids, bases and heat.
Specifications

Chemical Identifiers

28380-24-7
724.973
DANUORFCFTYTSZ-FBMOGUJMSA-N
123134611
CC1CCC(OC1C(C)C(=O)O)CC2CC(C(C3(O2)C(CC(O3)(C)C4CCC(O4)(C)C5C(CC(O5)C6C(CC(C(O6)(CO)O)C)C)C)C)C)OC
C40H68O11
MFCD00036825
nigericin
(2R)-2-[(2R,3S,6R)-6-[[(2S,4R,5R,6R,7R,9R)-2-[(2R,5S)-5-[(3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]-5-methyloxolan-2-yl]-7-methoxy-2,4,6-trimethyl-1,10-dioxaspiro[4.5]decan-9-yl]methyl]-3-methyloxan-2-

Specifications

White
28380-24-7
MFCD00036825
3892398
nigericin
DANUORFCFTYTSZ-FBMOGUJMSA-N
(2R)-2-[(2R,3S,6R)-6-[[(2S,4R,5R,6R,7R,9R)-2-[(2R,5S)-5-[(3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]-5-methyloxolan-2-yl]-7-methoxy-2,4,6-trimethyl-1,10-dioxaspiro[4.5]decan-9-yl]methyl]-3-methyloxan-2-
123134611
Powder
Nigericin sodium salt
10mg
C40H68O11
UN3462
14,6541
Soluble in chloroform, methanol or ethanol
CC1CCC(OC1C(C)C(=O)O)CC2CC(C(C3(O2)C(CC(O3)(C)C4CCC(O4)(C)C5C(CC(O5)C6C(CC(C(O6)(CO)O)C)C)C)C)C)OC
724.973
746.94
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

RTECSNumber : QT6840000

Recommended Storage : 2° to 8°C