Please login to your online account to display your discounted pricing

NIDA 41020, Tocris™

High affinity CB1 antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material NIDA 41020
Assay Percent Range >99%
Molecular Formula C23H24Cl2N4O2
Formula Weight 459.37
Synonym N-(Piperidin-1-yl)-5-(4-methoxyphenyl)-1-(2, 4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

392110

 
r & d systems
392110
10mg Each for $239.63

392150

 
r & d systems
392150
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 459.37
Quantity 10mg
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Synonym N-(Piperidin-1-yl)-5-(4-methoxyphenyl)-1-(2, 4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide
Chemical Name or Material NIDA 41020
Molecular Formula C23H24Cl2N4O2
CAS 502486-89-7
Safety and Handling

Store at Room Temperature