Please login to your online account to display your discounted pricing

Nicotinoyl chloride hydrochloride, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338410250

 View more versions of this product

Catalog No. AC338410250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (RCOCl)
Physical Form Crystalline Powder
Formula Weight 178.02
Quantity 25g
Color White to off-white
EINECS Number 243-648-1
TSCA TSCA
Melting Point 151° to 156°C
CAS Min % 96.0
CAS Max % 100.0
Synonym Pyridine-3-carbonyl chloride hydrochloride
Chemical Name or Material Nicotinoyl chloride hydrochloride
Infrared Spectrum Authentic
Name Note 97%
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C6H4ClNO·HCl
MDL Number MFCD00012820
CAS 20260-53-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Nicotinoyl chloride hydrochloride
Name Note 97%
CAS 20260-53-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (RCOCl)
Assay Percent Range 97%
Molecular Formula C6H4ClNO·HCl
Formula Weight 178.02
MDL Number MFCD00012820
Synonym Pyridine-3-carbonyl chloride hydrochloride
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents